Key Documents
BMSIPD960025TC
Sigma-Aldrich
BeadBlaster 96 Grinding Jars for Very Hard Samples
tungsten carbide, capacity 25 mL, pack of 2
About This Item
UNSPSC Code:
41100000
NACRES:
NA.82
material
tungsten carbide
packaging
pack of 2
manufacturer/tradename
Benchmark Scientific IPD9600-25TC
capacity
25 mL
compatibility
BeadBlaster 96
Related Categories
General description
Very hard samples require a jar and grinding balls that are just as hard or harder for effective sample preparation. All of the jars feature threaded closures for leakproof performance with both solids and liquids.
Tungsten carbide is an exceptionally hard ceramic material. It is useful for grinding materials where the presence of trace metals may be a concern. Applications include preparation of samples for XRF and particle reduction of ores, minerals, metal oxides and cement clinkers. These jars are extremely durable and offer a long service life.
Legal Information
BeadBlaster is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)