Skip to Content
Merck

Skip To

B7035

Nalgene® bottles, style 2125

capacity 2,000 mL

Synonym(s):

HDPE bottles, nalgene bottle, nalgene bottles, Nalgene 2125-2000, laboratory container, vessel

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NB.17
UNSPSC Code:
41121800
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

high-density polyethylene bottle, natural bottle, polypropylene cap, round bottle, white polypropylene screw closure, wide-mouth bottle

joint

Joints threaded neck (53 B)

description

Not autoclavable

feature

autoclavable: no, with cap

packaging

pack of 2 ea

manufacturer/tradename

Nalgene® 2125-2000

bottle diam. × H

119 mm × 260 mm

capacity

2,000 mL

General description

Round, heavy-duty, thick walled, wide-mouth bottles.

Features and Benefits

  • Nalgene® wide mouth heavy-duty bottles are specially designed for extra-rugged use.
  • Ideal for storing and transporting liquids or dry chemicals.
  • Includes leak-proof closure for preventing sample loss.
  • Suitable for use as a reservoir and waste trap in automated instrumentation.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B7160B0283B6910
material

high-density polyethylene bottle, polypropylene cap, white polypropylene screw closure, natural bottle, wide-mouth bottle, round bottle

material

high-density polyethylene bottle, polypropylene cap, round natural bottle, white polypropylene screw closure

material

polypropylene cap, polypropylene copolymer bottle, polypropylene copolymer bottles (PPCO), round bottle

material

high-density polyethylene bottle, polypropylene cap, round natural bottle, white polypropylene screw closure

manufacturer/tradename

Nalgene® 2125-2000

manufacturer/tradename

Nalgene® 2125-4000

manufacturer/tradename

Nalgene® 2105-0032

manufacturer/tradename

Nalgene® 2125-1000

feature

autoclavable: no, with cap

feature

with cap

feature

with cap

feature

with cap

capacity

2,000 mL

capacity

4,000 mL

capacity

1,000 mL

capacity

1,000 mL

joint

Joints threaded neck (53 B)

joint

Joints threaded neck (83B)

joint

Joints threaded neck (63)

joint

Joints threaded neck (538)

packaging

pack of 2 ea

packaging

pack of 1 ea

packaging

pack of 6 ea

packaging

pack of 6 ea


Still not finding the right product?

Explore all of our products under Nalgene® bottles, style 2125




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library