Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.17
UNSPSC Code:
41121800
material
high-density polyethylene bottle, polypropylene cap, polypropylene screw closure, square translucent bottle
joint
Joints threaded neck (63)
description
Not autoclavable
feature
with cap
packaging
pack of 6 ea
manufacturer/tradename
Nalgene® 2114-0032
bottle L × H × W
95 mm × 181 mm × 95 mm
capacity
1,000 mL
General description
Easily fill square wide-mouth bottles. HDPE exhibits excellent chemical resistance to most acids, bases and alcohols, making these bottles suitable for a wide variety of uses. Good for freezer storage to -100°C (-148°F). This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.
Features and Benefits
- Wide mouth is easy to fill.
- Bottle design saves space in freezer.
- HDPE exhibits excellent chemical resistance to most acids, bases and alcohols, making these bottles suitable for a wide variety of uses.
- Linerless caps provide leakproof† protection without the use of a liner that can wrinkle, cause leaks, or contaminate your reagents.
- Leakproof† when used with Nalgene closures.
- Bottle and Carton are readily recyclable in most communities.
- Manufactured from non-toxic materials.
- Participates in My Green Lab′s ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Still not finding the right product?
Explore all of our products under Nalgene® bottles, style 2114
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| B6785-6EA | 04061832613727 |