Skip to Content
Merck
All Photos(3)

Documents

AV47410

Sigma-Aldrich

Anti-TMEM30A antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-C6orf67, Anti-CDC50A, Anti-FLJ10856, Anti-Transmembrane protein 30A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

41 kDa

species reactivity

human, dog, guinea pig, horse, rat, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TMEM30A(55754)

General description

TMEM30A codes for a transmembrane protein that is present in the endoplasmic reticulum. Studies have reported that TMEM30A facilitates the import of bioactive and anticancer phospholipids into mammalian cells.
Rabbit Anti-TMEM30A recognizes human, mouse, rat, zebrafish, chicken, canine, and bovine TMEM30A.

Immunogen

Synthetic peptide directed towards the middle region of Human TMEM30A

Application

Rabbit Anti-TMEM30A antibody is suitable for western blot applications at a concentration of 0.25 μg/ml and for immunohistochemistry at 4-8 μg/ml.

Biochem/physiol Actions

The function of this protein remains unknown.

Sequence

Synthetic peptide located within the following region: FTLEKSFEGNVFMYYGLSNFYQNHRRYVKSRDDSQLNGDSSALLNPSKEC

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rui Chen et al.
Journal of immunology (Baltimore, Md. : 1950), 186(5), 3215-3225 (2011-02-04)
Antitumor alkylphospholipids initiate apoptosis in transformed HL-60 and Jurkat cells while sparing their progenitors. 1-O-Alkyl-2-carboxymethyl-sn-glycero-3-phosphocholine (Edelfosine) like other short-chained phospholipids--inflammatory platelet-activating factor (PAF) and apoptotic oxidatively truncated phospholipids--are proposed to have intracellular sites of action, yet a conduit for these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service