Skip to Content
Merck
All Photos(1)

Key Documents

32857

Sigma-Aldrich

Wright Stain solution

according to Wright

Synonym(s):

Eosin Methylene blue solution according to Wright, Wright’s eosin methylene blue

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
MA.02

form

liquid

Quality Level

quality

according to Wright

impurities

methanol

density

0.8 g/cm3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[K+].COC(=O)c1ccccc1C2=C3C=C(Br)C(=O)C(Br)=C3Oc4c(Br)c([O-])c(Br)cc24

InChI

1S/C21H10Br4O5.K/c1-29-21(28)9-5-3-2-4-8(9)14-10-6-12(22)17(26)15(24)19(10)30-20-11(14)7-13(23)18(27)16(20)25;/h2-7,26H,1H3;/q;+1/p-1

InChI key

NXBZXYJXNIFCRJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Wright′s stain solution is commercially available and is generally used in hematology. The procedure of making it includes heating methylene blue with sodium bicarbonate, followed by combining the mixture with eosin. It is an alkaline solution; hence its concentration is continuously changing, owing to the progressive oxidizing action of the alkali, therefore Giemsa′s stain is added to preserve the azurophilic properties of the polychrome dyes.

Application

Wright stain solution was used during the exudate analysis of white blood cell concentration.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simplified Wright's stain technique for routine avian blood smear staining.
Shen PF and Patterson LT
Poultry Science, 62 (5), 923-924 (1983)
Biocompatibility and biofouling of MEMS drug delivery devices.
Biomaterials, 24 (11), 1959-1967 (2003)
Qian Kong et al.
Microvascular research, 130, 104009-104009 (2020-04-26)
The purpose of the present study was to investigate the possible role of TIPE2 on acute lung injury (ALI) induced by myocardial ischemia/reperfusion (MIR) in diabetic rats. Sprague-Dawley (SD) rats were randomly separated into four groups: control+sham (C + sham); control+MIR (C + MIR);
Young-Joo Jeong et al.
Anatomy & cell biology, 43(4), 294-302 (2011-01-27)
Asthma is a Th2-dependent disease mediated by IgE and Th2 cytokines, and asthmatic patients suffer from oxidative stresses from abnormal airway inflammation. Vitamin C is a micro-nutrient functioning as an antioxidant. When administered at a mega-dose, vitamin C has been
Qian Kong et al.
Experimental and therapeutic medicine, 17(5), 3598-3606 (2019-04-17)
Acute lung injury (ALI) is a critical syndrome that is associated with high morbidity and mortality rates. The activation of the Fas/Fas ligand (FasL) signaling pathway may be an important pathophysiological mechanism during ALI development. Penehyclidine hydrochloride (PHC) has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service