T17917
Tricine
≥99% (titration), reagent grade, crystalline
Synonym(s):
N-[Tris(hydroxymethyl)methyl]glycine
About This Item
Recommended Products
biological source
synthetic
Quality Level
grade
reagent grade
Assay
≥99% (titration)
form
crystalline
storage condition
(Tightly closed. Dry)
impurities
<0.5% water
color
white
useful pH range
7.4-8.8
pKa (25 °C)
8.1
mp
187 °C
solubility
water: 0.1 M, clear, colorless
cation traces
Pb: ≤5 ppm
storage temp.
room temp
SMILES string
OCC(CO)(CO)NCC(O)=O
InChI
1S/C6H13NO5/c8-2-6(3-9,4-10)7-1-5(11)12/h7-10H,1-4H2,(H,11,12)
InChI key
SEQKRHFRPICQDD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Tricine′s valuable properties for biological research include its zwitterionic nature, wide buffering range (pH 7.4-8.8), classification as a Good′s buffering agent, non-toxicity to cells, and high water solubility and clarity. These attributes make it suitable for metal-sensitive systems, stable pH maintenance, optimal biological applications, cell biology studies, and accurate measurements. Additionally, Tricine may find utilization in capillary zone electrophoresis, high-performance liquid chromatography, and ion exchange chromatography, demonstrating adaptability across analytical techniques.
Application
- Electrophoresis To separate low-weight peptides and proteins
- Crystallization of proteins of varying pH
- Resuspension of cell pellets
- Enzyme assays
- Tissue culture
Features and Benefits
- Ideal as a Biological buffer for Cell Biology and Biochemical research
- Effective Buffering from pH 7.4 - 8.8 with a pKa of 8.1 (25 °C)
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service