PHR2475
Guaiacol Impurity E
Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Methyl benzoate
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
vapor density
4.68 (vs air)
vapor pressure
<1 mmHg ( 20 °C)
API family
guaiacol
form
liquid
packaging
pkg of 100 mg
refractive index
n20/D 1.516 (lit.)
bp
198-199 °C (lit.)
mp
−12 °C (lit.)
density
1.088 g/mL at 20 °C (lit.)
application(s)
pharmaceutical small molecule
storage temp.
2-8°C
SMILES string
COC(=O)c1ccccc1
InChI
1S/C8H8O2/c1-10-8(9)7-5-3-2-4-6-7/h2-6H,1H3
InChI key
QPJVMBTYPHYUOC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Other Notes
Footnote
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Repr. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
170.6 °F - Pensky-Martens closed cup
Flash Point(C)
77 °C - Pensky-Martens closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service