Skip to Content
Merck
All Photos(1)

Key Documents

PHR1596

Supelco

Pemetrexed

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Pemetrexed disodium heptahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19N5Na2O6 · 7H2O
CAS Number:
Molecular Weight:
597.48
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0001539

API family

pemetrexed

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[nH]1c2[nH]cc(c2[c](nc1N)=O)CCc3ccc(cc3)C(=O)N[C@@H](CCC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.O

InChI

1S/C20H21N5O6.2Na.7H2O/c21-20-24-16-15(18(29)25-20)12(9-22-16)6-3-10-1-4-11(5-2-10)17(28)23-13(19(30)31)7-8-14(26)27;;;;;;;;;/h1-2,4-5,9,13H,3,6-8H2,(H,23,28)(H,26,27)(H,30,31)(H4,21,22,24,25,29);;;7*1H2/q;2*+1;;;;;;;/p-2/t13-;;;;;;;;;/m0........./s1

InChI key

QJVSMHJWAOSBMD-MYXYZBIASA-L

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Pemetrexed is an anionic anticancer drug with a narrow therapeutic index. As a folate anti metabolite, it acts as an inhibitor of thymidylate synthase. It can act effectively against a broad-spectrum of solid tumors such as non-small-cell lung and breast cancers in clinical trials. It is also effective against both dihydrofolate reductase and glycinamide ribonucleotide formyl transferase.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA4712 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Muta. 2 - Repr. 1A - Skin Irrit. 2 - STOT RE 1

Target Organs

Blood,Immune system,Testes

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Utility of Schiff?s base Formation Reactions for the Spectrophotometric Determination of Pemetrexed Disodium in bulk and in its Pharmaceutical Formulations.
Khan PS, et al.
Asian Journal of Research in Chemistry, 7(3), 323-326 (2014)
Maria M Posada et al.
Drug metabolism and disposition: the biological fate of chemicals, 43(3), 325-334 (2014-12-17)
Pemetrexed, an anionic anticancer drug with a narrow therapeutic index, is eliminated mainly by active renal tubular secretion. The in vitro to in vivo extrapolation approach used in this work was developed to predict possible drug-drug interactions (DDIs) that may

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service