PHL80182
Urushiol (15:3)
phyproof® Reference Substance
Synonym(s):
3-(8Z,11Z,14-Pentadecatrienyl)-1,2-benzenediol
About This Item
Recommended Products
grade
primary reference standard
product line
phyproof® Reference Substance
Assay
≥85.0% (HPLC)
manufacturer/tradename
PhytoLab
application(s)
food and beverages
format
neat
storage temp.
−20°C
SMILES string
Oc1c(cccc1CCCCCCC\C=C/C\C=C/CC=C)O
InChI
1S/C21H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-16-19-17-15-18-20(22)21(19)23/h2,4-5,7-8,15,17-18,22-23H,1,3,6,9-14,16H2/b5-4-,8-7-
Related Categories
General description
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service