Skip to Content
Merck

N2

General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified

viscosity 2.144 mPa.s (25 °C)

Synonym(s):

UKAS ISO/IEC17025 Certified Viscosity Std.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

viscosity

2.144 mPa.s(25 °C)

application(s)

cleaning products
cosmetics
cosmetics
flavors and fragrances
food and beverages
personal care

format

mixture

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon General Viscosity Standards can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.


Still not finding the right product?


signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

10 - Combustible liquids

flash_point_f

168.8 °F

flash_point_c

76 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library