Recommended Products
product line
Selectophore™
Quality Level
form
liquid
composition
Chloride ionophore, 5.0 wt. % (24897)
1-Decanol, 4.0 wt. % (30608)
2-Nitrophenyl octyl ether, 90.0 wt. %
Tetradodecylammonium tetrakis(4-chlorophenyl)borate, 1.0 wt. % (87255)
General description
Visit our Sensor Applications portal to learn more.
Application
Selected application examples
Packaging
Clear borosilicate glass, high hydrolytic resistance (Type I)
Legal Information
Selectophore is a trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
>230.0 °F - closed cup
Flash Point(C)
> 110 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Frontiers in plant science, 11, 955-955 (2020-07-17)
The concentration of CO2 in the atmosphere has increased over the past 200 years and is expected to continue rising in the next 50 years at a rate of 3 ppm·year-1. This increase has led to a decrease in seawater
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service