Skip to Content
Merck
All Photos(1)

Key Documents

94950

Supelco

4-Vinyl-1-cyclohexene

analytical standard

Synonym(s):

4-Ethenyl-1-cyclohexene, NSC 15760

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H9CH=CH2
CAS Number:
Molecular Weight:
108.18
Beilstein:
1901553
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.76 (vs air)

vapor pressure

10.2 mmHg ( 25 °C)

Assay

≥99.5% (GC)

autoignition temp.

517 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.463 (lit.)
n20/D 1.464

bp

126-127 °C (lit.)

mp

−101 °C (lit.)

density

0.831 g/mL at 20 °C
0.831 g/mL at 20 °C
0.832 g/mL at 25 °C (lit.)

application(s)

environmental
petroleum

format

neat

storage temp.

2-8°C

SMILES string

C=CC1CCC=CC1

InChI

1S/C8H12/c1-2-8-6-4-3-5-7-8/h2-4,8H,1,5-7H2

InChI key

BBDKZWKEPDTENS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Vinyl-1-cyclohexene can be synthesized via cyclo-dimerization of butadiene in the presence of nitrosyl iron catalysts like [Fe(NO)2(CO)2] or [{FeCl(NO)2}2]/reductant.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inorganic Reactions and Methods, Reactions Catalyzed by Inorganic Compounds (2009)
Gold(I)-catalyzed asymmetric cycloisomerization of eneallenes into vinylcyclohexenes.
Michael A Tarselli et al.
Angewandte Chemie (International ed. in English), 46(35), 6670-6673 (2007-07-31)
D A Keller et al.
Toxicology and applied pharmacology, 144(1), 36-44 (1997-05-01)
4-Vinylcyclohexene (4-VCH), the dimer of 1,3-butadiene, is an ovarian toxicant in mice due to the formation of a diepoxide metabolite, but the tissue-specific site of formation of the metabolites is unknown. Microsomal preparations from liver, lung, and ovaries obtained from
S M Fontaine et al.
Drug metabolism and disposition: the biological fate of chemicals, 29(9), 1236-1242 (2001-08-15)
4-Vinyl-1-cyclohexene (VCH) is ovotoxic in B6C3F(1) mice but not in Fischer-344 rats, which can be partially attributed to greater formation of toxic epoxides from VCH in mice compared with rats. Since repeated exposure to VCH is necessary to cause ovotoxicity
S M Borman et al.
Toxicology and applied pharmacology, 167(3), 191-198 (2000-09-14)
Extensive destruction of primordial follicles by exposure to ovarian toxicants can cause early menopause in women. Primordial follicle destruction is known to result from dosing of mice and rats with three polycyclic aromatic hydrocarbons (PAHs), contaminants commonly found in cigarette

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service