81506
Poly(methyl methacrylate) standard ReadyCal set Mp ~800-2′200′000
for GPC
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
for GPC
shelf life
limited shelf life, expiry date on the label
technique(s)
gel permeation chromatography (GPC): suitable
format
neat
storage temp.
2-8°C
Packaging
1 set contains 30 vials for 10 complete calibrations
Components
ReadyCal Standards are polymer cocktails pre-weighed into autosampler vials for the convenient calibration of GPC/SEC/GFC systems.
This kit contains 10 sets of 3 color-coded vials, each containing four polymethyl methacrylates of different molar masses. Molar masses range from ~800 to ~2,200,000 Da and are carefully selected so that polymer traces are baseline separated.
Each kit contains sufficient material for at least 10 calibrations and includes an instruction manual and calibration curve with measurement conditions.
Suitable for calibrations in THF, toluene, HFIP, acetone, DMF, DMAc, DMSO, chloroform and NMP.
This kit contains 10 sets of 3 color-coded vials, each containing four polymethyl methacrylates of different molar masses. Molar masses range from ~800 to ~2,200,000 Da and are carefully selected so that polymer traces are baseline separated.
Each kit contains sufficient material for at least 10 calibrations and includes an instruction manual and calibration curve with measurement conditions.
Suitable for calibrations in THF, toluene, HFIP, acetone, DMF, DMAc, DMSO, chloroform and NMP.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Iliano V Coutinho-Abreu et al.
mSphere, 5(5) (2020-09-11)
The major surface lipophosphoglycan (LPG) of Leishmania parasites is critical to vector competence in restrictive sand fly vectors in mediating Leishmania attachment to the midgut epithelium, considered essential to parasite survival and development. However, the relevance of LPG for sand
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service