Skip to Content
Merck
All Photos(1)

Key Documents

60489-M

Sigma-Aldrich

Pentane

≥99% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH3
CAS Number:
Molecular Weight:
72.15
Beilstein:
969132
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.21

vapor density

2.48 (vs air)

Assay

≥99% (GC)

form

liquid

autoignition temp.

500 °F

expl. lim.

~8.3 %

refractive index

n20/D 1.358 (lit.)

bp

35-36 °C (lit.)

mp

−130 °C (lit.)

density

0.626 g/mL at 25 °C (lit.)

SMILES string

CCCCC

InChI

1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3

InChI key

OFBQJSOFQDEBGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pentane (n-pentane) is a hydrocarbon. Its hydroisomerization using a nanocrystalline beta (BEA) zeolite has been proposed. It has been reported as one of the products formed from the peroxidation of unsaturated fatty acid.

Other Notes

Pertains only in Germany: unversteuert

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-56.2 °F - closed cup

Flash Point(C)

-49.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James Clarke et al.
Nature nanotechnology, 4(4), 265-270 (2009-04-08)
A single-molecule method for sequencing DNA that does not require fluorescent labelling could reduce costs and increase sequencing speeds. An exonuclease enzyme might be used to cleave individual nucleotide molecules from the DNA, and when coupled to an appropriate detection
Michael Mayer et al.
Biophysical journal, 85(4), 2684-2695 (2003-09-26)
We present a straightforward, accessible method for the fabrication of micropores with diameters from 2 to 800 micro m in films of amorphous Teflon (Teflon AF). Pores with diameters </=40 micro m made it possible to record ion fluxes through

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service