Skip to Content
Merck
All Photos(1)

Key Documents

34956-M

Sigma-Aldrich

Pentane

suitable for HPLC, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH3
CAS Number:
Molecular Weight:
72.15
Beilstein:
969132
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21

grade

HPLC grade

vapor density

2.48 (vs air)

Assay

≥99.0%

form

liquid

autoignition temp.

500 °F

expl. lim.

~8.3 %

technique(s)

HPLC: suitable

impurities

≤0.0005% free acid (as CH3COOH)
≤0.0005% non-volatile matter
≤1 ppb fluorescence (quinine) at 254 nm
≤1 ppb fluorescence (quinine) at 365 nm
<0.005% water (Karl Fischer)

evapn. residue

<0.0005%

transmittance

200 nm, ≥20%
210 nm, ≥60%
220 nm, ≥85%
230 nm, ≥95%
250 nm, ≥99%

refractive index

n20/D 1.358 (lit.)

bp

35-36 °C (lit.)

mp

−130 °C (lit.)

solubility

water: partially soluble ( 0.038 g/l at 20 °C (68 °F))

density

0.626 g/mL at 25 °C (lit.)

λ

neat

UV absorption

λ: 200 nm Amax: ≤0.70
λ: 210 nm Amax: ≤0.20
λ: 220 nm Amax: ≤0.07
λ: 230 nm Amax: ≤0.02
λ: 250 nm Amax: ≤0.004

application(s)

general analytical

SMILES string

CCCCC

InChI

1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3

InChI key

OFBQJSOFQDEBGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pentane (n-pentane) is a hydrocarbon. Its hydroisomerization using a nanocrystalline beta (BEA) zeolite has been proposed. It has been reported as one of the products formed from the peroxidation of unsaturated fatty acid.

Application


  • Proton Affinities of Alkanes.: This study explores the proton affinities of various alkanes, including pentane, using mass spectrometry. The findings provide valuable data for understanding the reactivity and stability of alkanes in different chemical environments. (Fu et al., 2022).

  • Gas Digital Microfluidic Platform: Application to Highly Volatile Compound Preconcentration.: This paper presents a digital microfluidic platform designed for the preconcentration of highly volatile compounds, such as pentane. The platform significantly improves the detection limits and analytical performance in volatile organic compound analysis. (Enel et al., 2022).

  • Impact of Chemical and Physical Properties of Organic Solvents on the Gas and Hydrogen Formation during Laser Synthesis of Gold Nanoparticles.: This research examines how the chemical and physical properties of solvents, including pentane, affect gas and hydrogen formation during the laser synthesis of gold nanoparticles. The study provides insights into optimizing synthesis conditions for nanoparticle production. (Fromme et al., 2023).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-40.0 °F

Flash Point(C)

-40 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James Clarke et al.
Nature nanotechnology, 4(4), 265-270 (2009-04-08)
A single-molecule method for sequencing DNA that does not require fluorescent labelling could reduce costs and increase sequencing speeds. An exonuclease enzyme might be used to cleave individual nucleotide molecules from the DNA, and when coupled to an appropriate detection
Michael Mayer et al.
Biophysical journal, 85(4), 2684-2695 (2003-09-26)
We present a straightforward, accessible method for the fabrication of micropores with diameters from 2 to 800 micro m in films of amorphous Teflon (Teflon AF). Pores with diameters </=40 micro m made it possible to record ion fluxes through

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service