Skip to Content
Merck

32679

Cyanuric acid-13C3

PESTANAL®, analytical standard

Synonym(s):

1,3,5-Triazine-2,4,6-triol-13C3, 2,4,6-Trihydroxy-1,3,5-triazine-13C3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
13C3H3N3O3
CAS Number:
Molecular Weight:
132.05
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

mp

>360 °C

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

mass shift

M+3

SMILES string

O=[13C]1N[13C](=O)N[13C](=O)N1

InChI

1S/C3H3N3O3/c7-1-4-2(8)6-3(9)5-1/h(H3,4,5,6,7,8,9)/i1+1,2+1,3+1

InChI key

ZFSLODLOARCGLH-VMIGTVKRSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Cyanuric acid-13C3


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library