Skip to Content
Merck
All Photos(1)

Key Documents

06474

Supelco

Amberlite XAD1180N polymeric adsorbent

20-60 mesh

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
SB.52

product name

Amberlite XAD1180N, 20-60 mesh

technique(s)

LPLC: suitable

surface area

450 m2/g

loss

~65% loss on drying, 110 °C

matrix

styrene/divinylbenzene

matrix active group

polymer

particle size

20-60 mesh
350-600 μm

pore size

1.4 mL/g pore volume
300-400 Å mean pore size

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.

Application

Polymeric adsorbent for hydrophobic compounds: antibiotics, vitamins, amino acids, enzymes; for purification: gallium, myxovirescins, phosphoric acid, riboflavin salts.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service