Skip to Content
Merck

SLAA033

Millex-AA Filter Unit (Sterile)

pore size 0.8 μm, diam. 33 mm, mixed cellulose esters (MCE) membrane, hydrophilic, medical device

Synonym(s):

disposable syringe filter, mixed cellulose esters syringe filter, sterile syringe filter, syringe filter

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41104922
NACRES:
NB.24
eCl@ss:
32031690
Pore size:
0.8 μm pore size
Diam.:
33 mm
Sterility:
sterile; ethylene oxide treated
Material:
mixed cellulose esters (MCE) membrane, modified acrylic housing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

mixed cellulose esters (MCE) membrane, modified acrylic housing

sterility

sterile; ethylene oxide treated

product line

Millex

feature

CE compliant, holdup volume≤ 0.1 mL, hydrophilic, medical device

manufacturer/tradename

Millex

parameter

10.3 bar max. inlet pressure (150 psi), 45 °C max. temp.

diam.

33 mm

filtration area

4.52 cm2

inlet to outlet H

27 mm

volume

100 mL

matrix

MF-Millipore

pore size

0.8 μm pore size

fitting

female Luer-Lok® inlet, male Luer outlet slip

shipped in

ambient

storage temp.

room temp

General description

The Millex-AA Filter Unit (Sterile) is a medical device intended for use as syringe filters to clarify low volume solutions in direct patient care and pharmacy admixture applications.

Application

  • Clinical benefits: Removal of microorganisms, particles, precipitates, and undissolved powders larger than the membrane’s rated pore size from solutions for clinical use.
  • Hospital pharmacy: Clarification of small volumes of protein pharmaceuticals, diagnostic imaging agents, chemotherapeutics, aqueous solutions, or water during admixture preparation.
  • Direct patient care: Particulate removal from epidural and other liquid anaesthetics as well as from irrigation solutions used in ophthalmic, otic, and other surgical procedures.

Features and Benefits

  • Bidirectional support of the filter membrane
  • Removes microorganisms, particles, precipitates, and undissolved powders larger than the membrane′s rated pore size
  • Single-use, non-pyrogenic and non-toxic
  • mixed cellulose esters (MCE) membrane filter sealed in a modified acrylic copolymer (MMA) housing
  • Compatible with most aqueous solutions
  • CE marked

Other Notes

Devices are intended for medical and/or laboratory professional use only.

Legal Information

Luer-Lok is a registered trademark of Becton-Dickinson & Co.
MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany
Millex is a trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Millex-AA Filter Unit (Sterile)


pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Sol. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library