Skip to Content
Merck
All Photos(1)

Key Documents

MAB1681

Sigma-Aldrich

Anti-Vimentin Antibody, clone LN-6

culture supernatant, clone LN-6, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

LN-6, monoclonal

species reactivity

human, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)

isotype

IgM

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... VIM(7431)

General description

Vimentin is a class III intermediate filament protein (homopolymers) found in various non-epithelial cells including mesenchymal cells and fibroblasts. In many cells of mesenchymal origin, it is one of the most prominent phosphoproteins, especially during cell division when enhanced phosphorylation of vimentin corresponds with the reorganization of vimentin filaments.

Specificity

Reacts with human mesenchymal cells and derived sarcomas. Reacts with an epitope of the cytoskeletal protein vimentin but unlike other vimentin antibodies, does not react with human lymphoid cells or derived malignancies. Reactive with endothelium, muscle, fibroblasts, melanocytes, peripheral nerve, sertoli cells, kidney mesangial cells and tubules, osteoblasts and periosteum, non-hematopoietic tumors of mesenchymal origin (all sarcomas), meningiomas and melanomas.

Immunogen

Nuclei from adult human thymic biopsy specimens.

Application

Anti-Vimentin Antibody, clone LN-6 detects level of Vimentin & has been published & validated for use in IH(P).
Immunohistochemistry on B5 fixed, paraffin embedded tissue sections.

Target description

57-60 kDa

Physical form

Liquid tissue culture supernatant containing 0.02% sodium azide.

Analysis Note

Control
Hyperplastic lymph nodes.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paola Valdivieso et al.
PloS one, 12(4), e0174864-e0174864 (2017-04-07)
Mechanical stress, including blood pressure related factors, up-regulate expression of the pro-angiogenic extracellular matrix protein tenascin-C in skeletal muscle. We hypothesized that increased capillarization of skeletal muscle with the repeated augmentation in perfusion during endurance training is associated with blood
Liqing Wang et al.
The Journal of comparative neurology, 524(11), 2322-2334 (2016-01-01)
Expression of Nogo protein was investigated in the optic pathway of embryonic mice by using isoform-specific antibodies Bianca and 11C7, which recognize Nogo-A/B and Nogo-A, respectively. Our previous reports from using antibody N18 have shown that Nogo is localized on
Francesca Cialdai et al.
Cellular and molecular life sciences : CMLS, 78(23), 7795-7812 (2021-10-30)
Astronauts on board the International Space Station (ISS) are exposed to the damaging effects of microgravity and cosmic radiation. One of the most critical and sensitive districts of an organism is the eye, particularly the retina, and > 50% of astronauts develop
David P J Hunt et al.
Stem cells (Dayton, Ohio), 26(1), 163-172 (2007-09-29)
Skin-derived precursor cells (SKPs) are multipotent neural crest-related stem cells that grow as self-renewing spheres and are capable of generating neurons and myelinating glial cells. SKPs are of clinical interest because they are accessible and potentially autologous. However, although spheres
Maria Elena Melica et al.
Stem cell research & therapy, 15(1), 20-20 (2024-01-18)
The glomerulus is a highly complex system, composed of different interdependent cell types that are subjected to various mechanical stimuli. These stimuli regulate multiple cellular functions, and changes in these functions may contribute to tissue damage and disease progression. To

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service