Skip to Content
Merck

Skip To

GF304

GM-CSF Protein, Human Recombinant Animal Free

The GM-CSF protein was initially characterized as a factor that can support the in vitro colony formation of granulocyte macrophage progenitors. The Recombinant animal free human GM-CSF is manufactured using all non-animal reagents.

Synonym(s):

GMCSF, M-CSF, CSF-1, Granulocyte Colony Stimulating Factor, Macrophage Colony Stimulating Factor, Granulocyte/Macrophage Colony Stimulating Factor, G-CSF

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
eCl@ss:
32160405
Assay:
>98% (SDS-PAGE and HPLC)
Recombinant:
expressed in E. coli
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in E. coli

Quality Segment

assay

>98% (SDS-PAGE and HPLC)

impurities

<0.1 μg/mg protein endotoxin (<1EU/μg)

UniProt accession no.

shipped in

dry ice

storage temp.

-10 to -25°C

General description

Granulocyte/Macrophage Colony-Stimulating Factor (GM-CSF) was initially characterized as a factor that can support the in vitro colony formation of granulocyte macrophage progenitors. It is also a growth factor for erythroid, megakaryocyte, and eosinophil progenitors. GMCSF is produced by a number of different cell types (including T cells, B cells, macrophages, mast cells, endothelial cells, fibroblasts, and adipocytes) in response to cytokine or inflammatory stimuli. The human and murine molecules are species-specific and exhibit no cross-species reactivity. Recombinant human GM-CSF is a 14.6 kDa globular protein consisting of 128 amino acids containing two intramolecular disulfide bonds and two potential N-linked glycosylation sites.
Product Source: E. coli

Application

Research Category
Inflammation & Immunology
Research Sub Category
Growth Factors & Receptors

Biochem/physiol Actions

Cross Reactivty
None

Physical form

Product is filtered through a 0.2 micron filter before lyophilization.
Recombinant animal free human GM-CSF is manufactured using all non-animal reagents

Preparation Note

Store at -20°C for up to 6 months from date of receipt.

Analysis Note

Bioactivity assay: The ED50 was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 107 units/mg.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
G5035SRP3110SRP3050
description

The GM-CSF protein was initially characterized as a factor that can support the in vitro colony formation of granulocyte macrophage progenitors. The Recombinant animal free human GM-CSF is manufactured using all non-animal reagents.

description

GM-CSF, recombinant, expressed in E. coli, suitable for cell culture

description

Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

description

GM-CSF, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture

recombinant

expressed in E. coli

recombinant

expressed in E. coli

recombinant

expressed in E. coli

recombinant

expressed in E. coli

assay

>98% (SDS-PAGE and HPLC)

assay

≥98% (SDS-PAGE)

assay

≥98% (SDS-PAGE), ≥98% (HPLC)

assay

≥98% (SDS-PAGE)

impurities

<0.1 μg/mg protein endotoxin (<1EU/μg)

impurities

≤1.000 EU/μg

impurities

<0.1 EU/μg endotoxin, tested

impurities

≤1 EU/μg endotoxin, tested

UniProt accession no.

P09603

UniProt accession no.

P04141

UniProt accession no.

P09603

UniProt accession no.

P04141

storage temp.

-10 to -25°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

shipped in

dry ice

shipped in

-

shipped in

wet ice

shipped in

wet ice


Storage Class

11 - Combustible Solids

WGK

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters