Skip to Content
Merck
All Photos(2)

Key Documents

CKT2MAG-97K

Millipore

MILLIPLEX® Canine Kidney Toxicity Panel 2 - Toxicity Multiplex Assay

The analytes available for this multiplex kit are: Albumin, β-2-microglobulin (B2M), Trefoil Factor 3 (TFF-3), Retinol Binding Protein 4 (RBP4)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity

canine

manufacturer/tradename

Milliplex®

assay range

accuracy: 96-102%
sensitivity: 0.016-0.026 ng/mL
(All other analytes; MinDC +2SD)

sensitivity: 12.48 ng/mL
(Albumin; MinDC +2SD)

standard curve range: 0.01-50 ng/mL
(RBP4)

standard curve range: 0.02-100 ng/mL
(B2M)

standard curve range: 0.02-100 ng/mL
(TFF-3)

standard curve range: 12.21-50,000 ng/mL
(Albumin)

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

As one of the leading causes of drug candidate attrition, toxicity research is receiving more attention, especially in the search of sensitive, organ-specific toxicity biomarkers. In pharmaceutical and chemical industries, renal function is routinely assessed during toxicity evaluations. The importance of the kidney as a central detoxification organ leads to a high exposure of renal tissue to drugs, reactive metabolites, and environmental compounds. Traditional markers for assessing renal toxicity, such as blood urea nitrogen (BUN) and serum creatinine, are insensitive and non-specific. For improved, non-invasive detection of acute nephrotoxicity, a panel of novel urinary kidney biomarkers has been identified and validated for preclinical studies in relevant animal species.

MILLIPLEX® Canine Kidney Toxicity Panel 2 is a 4-plex kit to be used for the simultaneous quantification of any combination of the following analytes in canine urine samples: Albumin*, β-2-microglobulin (B2M)*, Trefoil Factor 3 (TFF-3)*, Retinol Binding Protein 4 (RBP4)*. This kit uses a 96-well format, contains a lyophilized standard cocktail, two internal assay quality controls and can measure up to 38 samples in duplicate.
*Urinary albumin, B2M, TFF-3 and RBP4 are biomarkers listed in the Predictive Safety Testing Consortium (PSTC) project pipeline which have a strong translational role in drug safety testing.

The Luminex® xMAP® platform uses a magnetic bead immunoassay format for ideal speed and sensitivity to quantitate multiple analytes simultaneously, dramatically improving productivity while conserving valuable sample volume.

Panel Type: Toxicity

Specificity

Cross Reactivty
Approximately 5% cross-reactivity may be observed between B2M beads and TFF-3 protein. No other significant cross-reactivity was observed for other analytes in this panel.

Application

  • Analytes: Albumin, β-2-Microglobulin (B2M), Trefoil Factor 3 (TFF-3), Retinol Binding Protein 4 (RBP4)
  • Recommended Sample Type: Canine urine
  • Recommended Sample Dilution: 25 μL of 1:500 diluted canine urine for normal subjects; optimal dilutions for disease subjects should be determined
  • Assay Run Time: Primary incubation of 2 hours, followed by a secondary incubation of 1 hour, both at room temperature (20-25°C)
  • Research Category: Toxicology

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Components

Canine Kidney Toxicity Panel 2 Standard, 1 vial, lyophilized

Canine Kidney Toxicity Panel 2 Control 1, 1 vial, lyophilized

Canine Kidney Toxicity Panel 2 Control 2, 1 vial, lyophilized

Assay Buffer, 2 bottles, 30 mL

Wash Buffer, 10X (0.05% Proclin), 2 bottlesl, 30 mL

Canine Kidney Toxicity Panel 2 Detection Antibodies, 1 bottle, 5.5 mL

Streptavidin-Phycoerythrin, 1 bottle, 5.5 mL

Mixing Bottle, 1 bottle

Sample Plate, 96 well format, 1 plate

2 Foil Plate Sealers

Other Notes

Please contact Technical Service for linearity of dilution.
Sensitivity: See kit protocol for individual analyte sensitivities.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Novel kidney toxicity biomarkers expand options for acute nephrotoxicity detection. Multiplex assays measure multiple renal damage biomarkers in small sample volumes, minimizing time and costs. See how MILLIPLEX® multiplex kidney toxicity assays detected vancomycin-induced subacute nephrotoxicity in rat models.

Novel kidney toxicity biomarkers expand options for acute nephrotoxicity detection. Multiplex assays measure multiple renal damage biomarkers in small sample volumes, minimizing time and costs. See how MILLIPLEX® multiplex kidney toxicity assays detected vancomycin-induced subacute nephrotoxicity in rat models.

Novel kidney toxicity biomarkers expand options for acute nephrotoxicity detection. Multiplex assays measure multiple renal damage biomarkers in small sample volumes, minimizing time and costs. See how MILLIPLEX® multiplex kidney toxicity assays detected vancomycin-induced subacute nephrotoxicity in rat models.

Novel kidney toxicity biomarkers expand options for acute nephrotoxicity detection. Multiplex assays measure multiple renal damage biomarkers in small sample volumes, minimizing time and costs. See how MILLIPLEX® multiplex kidney toxicity assays detected vancomycin-induced subacute nephrotoxicity in rat models.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service