Skip to Content

AP162F

Rabbit Anti-Chicken IgG Antibody, FITC conjugate

1.5 mg/mL, Chemicon®

Synonym(s):

IgY

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
eCl@ss:
32160702
Conjugate:
FITC conjugate
Clone:
polyclonal
Application:
IF
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

FITC conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

chicken

manufacturer/tradename

Chemicon®

concentration

1.5 mg/mL

technique(s)

immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

Application

Detect Chicken IgG using this Rabbit anti-Chicken IgG Antibody, FITC conjugate validated for use in IF.
Suggested dilution for most applications: 1:50-1:200 for most applicationsOptimal working dilutions must be determined by the end user.

Biochem/physiol Actions

Chicken IgY (IgG (H+L))

ANTIBODY SPEIFICITY:

Based on immunoelectrophoresis, the antibody reacts with the heavy chainjs of Chicken IgY and with light chains common to most chicken immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins, but antibodies may cross-react with immunoglobulins from other species.

FLUOROPHORE /PROTEIN RATIO

7.0 ug/mg; 2.6 moles FITCper mole IgG.

Physical form

Lyophilized. Buffer = 0.01 M Sodium Phosphate, 0.25 M NaCl, pH 7.6. Preservative 0.01% Thimerosal, 0.05% Sodium Azide with 15 mg/mL BSA.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Try our Product Selector Tool to narrow your options


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 3

Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library