Skip to Content
Merck
All Photos(2)

Documents

ABT338

Sigma-Aldrich

Anti-EBI3 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Interleukin-27 subunit beta, IL-27 subunit beta, IL-27B, Epstein-Barr virus-induced gene 3 protein, EBV-induced gene 3 protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... EBI3(10148)

Related Categories

General description

Epstein-Barr virus-induced gene 3 protein (EBI3), also known as Interleukin-27 subunit beta (IL27B), is a member of the type I cytokine receptor family (Type 3 subfamily) that is composed of two subunits: interleukin-27 (IL-27) and IL-35. EBI3 regulates T-helper cell development, stimulates cytotoxic T-cell activity, suppresses T-cell proliferation, and induces isotype switching in B-cells. EBI3 also plays an important role in its antitumor activity as well as its antiangiogenic activity.

Specificity

This antibody recognizes the C-terminus of EBI3.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of human EBI3.

Application

Detect IL-27 using this rabbit polyclonal antibody, Anti-EBI3 Antibody validated for use in western blotting, IHC & Immunofluorescence.
Immunohistochemistry Analysis: 2.5 µg/mL from a representative lot detected EBI3 in human spleen tissue.

Immunofluorescence Analysis: 20 μg/mL from a representative lot detected EBI3 in human spleen cells.
Research Category
Cell Structure
Research Sub Category
Cytokines & Cytokine Receptors

Quality

Evaluated by Western Blotting/Peptide Inhibition Assay in K562 cell lysate.

Western Blotting/Peptide Inhibition Assay Analysis: 1 µg/mL of this antibody detected EBI3 in 15 µg of K562 cell lysate.

Target description

~37 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
K562 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service