Sign In to View Organizational & Contract Pricing.
Select a Size
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IP, WB
Citations:
-
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
serum
antibody product type
primary antibodies
clone
polyclonal
species reactivity
rat, mouse, human
technique(s)
immunoprecipitation (IP): suitable, western blot: suitable
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
mouse ... Paxbp1(67367)
General description
In skeletal muscles, Pax7 uniquely marks muscle satellite cells and plays some important yet unknown functions at the perinatal stage. Pax3/7BP is a ubiquitously expressed nuclear protein, enriched in Pax7+ muscle precursor cells (MPCs), and serves as an indispensable adaptor for Pax7 to recruit the histone-3-lysine-4 (H3K4) methyltransferase (HMT) complex by bridging Pax7 and Wdr5. Recently, Wu et al. have established Pax3/7BP as an essential adaptor linking Pax3/7 with the H3K4 HMT to regulate the proliferation of muscle progenitor cells.
~105 kDa observed. Uncharacterized bands may be observed in some lysate(s).
Immunogen
Recombinant protein corresponding to mouse PAX3/7BP.
Application
Anti-PAX3/7BP Antibody is an antibody against PAX3/7BP for use in western blotting & IP.
Research Category
Stem Cell Research
Stem Cell Research
Research Sub Category
Muscle Physiology
Developmental Signaling
Muscle Physiology
Developmental Signaling
Physical form
Unpurified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Evaluated by Western Blot in a jurkat cell lysate.
Western Blot Analysis: A 1:1000 dilution of this antibody detected Pax3/7BP in a 10 µg jurkat cell lysate.
Western Blot Analysis: A 1:1000 dilution of this antibody detected Pax3/7BP in a 10 µg jurkat cell lysate.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| ABD85 | 04053252939563 |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service