Skip to Content
Merck
All Photos(1)

Key Documents

902282

Supelco

Rapid-action connector

for use with product tube (8 x 6 mm)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121815
NACRES:
NB.51

compatibility

for use with product tube (8 x 6 mm)

shipped in

ambient

storage temp.

no temp limit

Related Categories

General description

Our withdrawal and safety components are easily interconnectable thanks to a wide selection of reducers, adapters and couplings. Due to the wide variety, many different customized installations are possible – everything for your individual requirements. This rapid-action connector is made of stainless steel with a special high quality sealing for maximum protection of the our solvents quality. It is equipped with nipple (male) Type 21 and tube connector 8x6 mm.
for product tube (8 x 6 mm)

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service