8.22289
1-Naphthol
for synthesis
Synonym(s):
1-Naphthol, 1-Hydroxynaphthalene
About This Item
Recommended Products
vapor pressure
1.3 hPa ( 94 °C)
Quality Level
Assay
≥99% (GC)
form
solid
autoignition temp.
510 °C
potency
1870 mg/kg LD50, oral (Rat)
880 mg/kg LD50, skin (Rabbit)
technique(s)
UV/Vis spectroscopy: suitable
bp
288 °C/1013 hPa
mp
94-96 °C
transition temp
flash point 125 °C
solubility
0.1 g/L
density
1.28 g/cm3 at 20 °C
bulk density
450 kg/m3
storage temp.
2-30°C
SMILES string
Oc1c2c(ccc1)cccc2
InChI
1S/C10H8O/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7,11H
InChI key
KJCVRFUGPWSIIH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
<li><strong>Real-time deflection and friction force imaging by bimorph-based resonance-type high-speed scanning force microscopy in the contact mode.</strong>: This study employs fluorine-doped tin oxide coated glass slides in a novel microscopy technique that enhances real-time imaging capabilities, demonstrating its applicability in high-speed, high-resolution surface analysis (Cai et al., 2014).</li>
<li><strong>Langmuir-Blodgett films of self-assembled (alkylether-derivatized Zn phthalocyanine)-(C imidazole adduct) dyad with controlled intermolecular distance for photoelectrochemical studies.</strong>: This research explores the use of fluorine-doped tin oxide as a substrate for Langmuir-Blodgett films, focusing on their potential in creating highly organized molecular assemblies for enhanced photoelectrochemical applications (Obraztsov et al., 2014).</li>
<li><strong>Formation of an electron hole doped film in the α-Fe2O3 photoanode upon electrochemical oxidation.</strong>: The paper describes the advantageous characteristics of fluorine-doped tin oxide in forming electron hole doped films on α-Fe2O3 photoanodes, pivotal for improving the efficiency of photoelectrochemical cells (Gajda-Schrantz et al., 2013).</li>
<li><strong>BiSI Micro-Rod Thin Films: Efficient Solar Absorber Electrodes</strong>: Investigates the role of fluorine-doped tin oxide coated glass slides in enhancing the photoelectrochemical properties of BiSI micro-rod thin films, suggesting potential applications in solar energy conversion and storage systems (Hahn et al., 2012).</li>
</ul>
Analysis Note
Melting range (lower value): ≥ 94 °C
Melting range (upper value): ≤ 96 °C
Identity (IR): passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1A - STOT SE 2 Oral - STOT SE 3
Target Organs
Kidney, Respiratory system
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 1
Flash Point(F)
257.0 °F - closed cup
Flash Point(C)
125 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service