Skip to Content
Merck
All Photos(1)

Key Documents

8.21752

Sigma-Aldrich

N,N-Diethylformamide

for synthesis

Synonym(s):

N,N-Diethylformamide, Formic acid diethylamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCON(C2H5)2
CAS Number:
Molecular Weight:
101.15
MDL number:
UNSPSC Code:
12352111
EC Index Number:
210-533-2
NACRES:
NA.22

Quality Level

Assay

≥99.0% (GC)

form

liquid

transition temp

flash point 65 °C

density

0.91 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(CC)(CC)C=O

InChI

1S/C5H11NO/c1-3-6(4-2)5-7/h5H,3-4H2,1-2H3

InChI key

SUAKHGWARZSWIH-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.905 - 0.907
Water (K. F.): ≤ 0.20 %
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service