Skip to Content
Merck
All Photos(2)

Key Documents

8.16147

Sigma-Aldrich

(R)-(-)-Menthyl acetate

for synthesis

Synonym(s):

(R)-(-)-Menthyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O2
CAS Number:
Molecular Weight:
198.30
MDL number:
UNSPSC Code:
12352108
EC Index Number:
220-076-0
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC)

form

liquid

potency

>5000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

density

0.925 g/cm3

storage temp.

2-30°C

SMILES string

O([C@H]1[C@@H](CC[C@H](C1)C)C(C)C)C(=O)C

InChI

1S/C12H22O2/c1-8(2)11-6-5-9(3)7-12(11)14-10(4)13/h8-9,11-12H,5-7H2,1-4H3/t9-,11+,12-/m1/s1

InChI key

XHXUANMFYXWVNG-ADEWGFFLSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.923 - 0.928
Spec. rotation [α²0/D (c=8 in chloroform): -93.0 - -83.0 °
Identity (IR): passes test

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service