Skip to Content
Merck
All Photos(2)

Key Documents

8.04308

Sigma-Aldrich

1,6-Hexanediol

for synthesis

Synonym(s):

1,6-Hexanediol, Hexamethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2)6OH
CAS Number:
Molecular Weight:
118.17
MDL number:
UNSPSC Code:
12352104
EC Index Number:
211-074-0
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

Assay

≥96.5% (GC)

form

crystals

autoignition temp.

320 °C

potency

3730 mg/kg LD50, oral (Rat)

expl. lim.

6.6-16 % (v/v)

pH

7.6 (20 °C, 900 g/L in H2O)

bp

253-260 °C

mp

37-42 °C

transition temp

flash point 147 °C

solubility

5000 g/L

density

1.12 g/cm3 at 20 °C

bulk density

530 kg/m3

storage temp.

2-30°C

SMILES string

OCCCCCCO

InChI

1S/C6H14O2/c7-5-3-1-2-4-6-8/h7-8H,1-6H2

InChI key

XXMIOPMDWAUFGU-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.5 % (a/a)
Melting range (lower value): ≥ 37 °C
Melting range (upper value): ≤ 42 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service