Skip to Content
Merck
All Photos(1)

Key Documents

8.01986

Sigma-Aldrich

Bromotrichloromethane

for synthesis

Synonym(s):

Bromotrichloromethane, Trichlorobromomethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CBrCl3
CAS Number:
Molecular Weight:
198.27
MDL number:
UNSPSC Code:
12352101
EC Index Number:
200-886-0
NACRES:
NA.22

vapor pressure

49 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

reaction suitability

reaction type: C-C Bond Formation

bp

105 °C/1013 hPa

mp

-6 °C

density

2.01 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

BrC(Cl)(Cl)Cl

InChI

1S/CBrCl3/c2-1(3,4)5

InChI key

XNNQFQFUQLJSQT-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 2.005 - 2.015
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service