Skip to Content
Merck
All Photos(1)

Key Documents

420139

Sigma-Aldrich

JAK2 Inhibitor IV

The JAK2 Inhibitor IV, also referenced under CAS 1110502-30-1, controls the biological activity of JAK2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

JAK2 Inhibitor IV, 3-Amino-5-(N- tert-butylsulfonamido-4-phenyl)-indazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N4O2S
CAS Number:
Molecular Weight:
344.43
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

ethanol: 3 mg/mL
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(NC(C)(C)C)c1ccc(cc1)c2cc3c([nH]nc3N)cc2

InChI

1S/C17H20N4O2S/c1-17(2,3)21-24(22,23)13-7-4-11(5-8-13)12-6-9-15-14(10-12)16(18)20-19-15/h4-10,21H,1-3H3,(H3,18,19,20)

InChI key

KFJCXIOVAGJCKB-UHFFFAOYSA-N

General description

An aminoindazole compound that potently inhibits the activity of both the wild-type JAK2 and the constitutively active V617F mutant (IC50 = 78 and 206 nM, respectively) frequently found in patients with clonal polycythemia vera, essential thrombocytosis, and chronic idiopathic myelofibrosis, while exhibiting much reduced activity against JAK3 (IC50 = 2.93 µM).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Antonysamy, S., et al. 2008. Bioorg. Med. Chem. Lett.19, 279.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong Yu et al.
Neural regeneration research, 18(5), 1132-1138 (2022-10-19)
Inhibiting retinal neovascularization is the optimal strategy for the treatment of retina-related diseases, but there is currently no effective treatment for retinal neovascularization. P-element-induced wimpy testis (PIWI)-interacting RNA (piRNA) is a type of small non-coding RNA implicated in a variety
Md Al Nayem Chowdhury et al.
Cell death & disease, 13(7), 619-619 (2022-07-20)
Checkpoint kinase 2 (CHK2) plays an important role in safeguarding the mitotic progression, specifically the spindle assembly, though the mechanism of regulation remains poorly understood. Here, we identified a novel mitotic phosphorylation site on CHK2 Tyr156, and its responsible kinase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service