Skip to Content
Merck
All Photos(2)

Key Documents

1.46570

Millipore

Buffered NaCl Peptone Solution

bottle capacity 125 mL, bottle filling volume 90 mL, closure type, Blue crimp cap with 3 septa, pack of 10 bottles

Synonym(s):

Buffered Saline Peptone Water, NaCl Peptone Buffer

Sign Into View Organizational & Contract Pricing


About This Item

Agency

EP 2.6.1
JP 4.06
USP 71
71

Quality Level

sterility

sterile; autoclaved

form

liquid

feature

closure type Blue crimp cap with 3 septa
ready-to-use

packaging

pack of 10 bottles

technique(s)

direct inoculation: suitable
microbiological culture: suitable

bottle capacity

125 mL

bottle filling volume

90 mL

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

components

lecithin
tween

storage temp.

2-25°C

suitability

nonselective for

General description

Buffered NaCl Peptone Solution with LT (lecithin and Tween®80) has a low content of peptone, along with phosphate buffer and sodium chloride, to maintain the viability of microorganisms present in the test sample without supporting their growth.

Lecithin and Polysorbate 80, is effective against quaternary ammonium compounds, amphoteric surfactants, benzamidines, chlorhexidines and dequadin. Polysorbate 80 inactivates benzyl alcohol, carbanilides, dichlorbenzyl alcohols, benzoic acid, p-hydroxybenzoic acid and its esters, phenols, phenylethyl alcohols and PHB esters.

Application

Buffered NaCl Peptone Solution with LT (lecithin and Tween®80) is used for the microbial examination of non-sterile samples. It is suitable for dilution, sample preparation and sample washing.

Features and Benefits

  • Our ready-to-use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements.
  • Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility, and growth promotion tests.
  • Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy, and significantly reducing the risk of incorrect interpretation and false results.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service