Skip to Content
Merck

Skip To

1.04201

Glycine

GR for analysis

Synonym(s):

Gly, Aminoacetic acid, Aminoethanoic acid, Aminoacetic acid, Aminoethanoic acid, Glycocoll

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
NH2CH2COOH
CAS Number:
Molecular Weight:
75.07
UNSPSC Code:
12352209
NACRES:
NA.21
EC Index Number:
200-272-2
MDL number:
Beilstein/REAXYS Number:
635782
eCl@ss:
32160406
Assay:
≥99.7% (perchloric acid titration)
Technique(s):
inhibition assay: suitable
Vapor pressure:
0.0000171 Pa ( 25 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

0.0000171 Pa ( 25 °C)

Quality Segment

assay

≥99.7% (perchloric acid titration)

form

solid

potency

7930 mg/kg LD50, oral (Rat)

technique(s)

inhibition assay: suitable

pH

5.9-6.4 (20 °C, 50 g/L in H2O)

pKa (25 °C)

(1) 2.35, (2) 9.60, 2.35

mp

240 °C (dec.) (lit.)

solubility

soluble 250 g/L

density

1.161 g/cm3 at 20 °C

bulk density

920 kg/m3

storage temp.

2-30°C

SMILES string

NCC(O)=O

InChI

1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)

InChI key

DHMQDGOQFOQNFH-UHFFFAOYSA-N

Application


  • Gut microbiota-bile acid crosstalk and metabolic fatty liver in spotted seabass (Lateolabrax maculatus): The role of a cholesterol, taurine and glycine supplement.: This study investigates the use of glycine in dietary supplements to modulate gut microbiota and bile acid metabolism, which significantly impacts liver health and fat metabolism in marine species, providing insights into nutritional strategies for aquaculture (Song et al., 2024).

Biochem/physiol Actions

Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Analysis Note

Assay (perchloric acid titration): ≥ 99.7 %
Identity (IR-spectrum): passes test
pH (50 g/l CO₂-free water): 5.9 - 6.3
Chloride (Cl): ≤ 0.003 %
Sulfate (SO₄): ≤ 0.0025 %
Heavy metals (as Pb): ≤ 0.001 %
Cu (Copper): ≤ 0.0001 %
Fe (Iron): ≤ 0.0001 %
Pb (Lead): ≤ 0.0001 %
NH₄ (Ammonium): ≤ 0.02 %
Foreign amino acids: ≤ 0.1 %
Other ninhydrine positive substances: ≤ 0.1 %
In water insoluble matter: ≤ 0.003 %
Sulfated ash (600 °C): ≤ 0.05 %

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8160134810OP94119
description

GR for analysis

description

for synthesis

description

OmniPur® Grade, >= 98.5%, molecular biology grade

description

analytical standard, for nitrogen determination according to Kjeldahl method

assay

≥99.7% (perchloric acid titration)

assay

≥99.0% (perchloric acid titration)

assay

-

assay

≥99.0%

technique(s)

inhibition assay: suitable

technique(s)

inhibition assay: suitable

technique(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable, titration: suitable

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

100

solubility

soluble 250 g/L

solubility

-

solubility

-

solubility

1 M HCl: 75 mg/L, clear, colorless (2.252g in 30mL)

form

solid

form

solid

form

crystalline powder

form

-

mp

240 °C (dec.) (lit.)

mp

240 °C (dec.) (lit.)

mp

240 °C (dec.) (lit.)

mp

240 °C (dec.) (lit.)


Still not finding the right product?

Explore all of our products under Glycine


Storage Class

11 - Combustible Solids

flash_point_f

No data available

flash_point_c

No data available



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters