Skip to Content
Merck
All Photos(1)

Key Documents

06-502

Sigma-Aldrich

Anti-STAT2 Antibody, CT

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

manufacturer/tradename

Upstate®

technique(s)

electrophoretic mobility shift assay: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... STAT2(6773)

Specificity

STAT2

Immunogen

GST fusion protein containing amino acids 671-806 of human STAT2

Application

Anti-STAT2 Antibody, CT is a Rabbit Polyclonal Antibody for detection of STAT2 also known as Signal Transducer & Activator of Transcription 2 & has been tested in EMSA, IP, WB, ICC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Immunoblot Analysis: 0.5-2 μg/ml of this antibody detected STAT2 (113kDa) in Jurkat cell lysates.

Target description

113kDa

Physical form

0.07 M Tris-glycine, pH 7.4, 0.105 M NaCl, 0.035% sodium azide and 30% glycerol.
Format: Purified
Protein A chromatography

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Martin-Fernandez et al.
Cell reports, 31(6), 107633-107633 (2020-05-14)
Most monogenic disorders have a primary clinical presentation. Inherited ISG15 deficiency, however, has manifested with two distinct presentations to date: susceptibility to mycobacterial disease and intracranial calcifications from hypomorphic interferon-II (IFN-II) production and excessive IFN-I response, respectively. Accordingly, these patients
Alicia L Humlicek et al.
Journal of immunology (Baltimore, Md. : 1950), 178(10), 6395-6403 (2007-05-04)
Respiratory pathogens and toxins often assault the lung from the airway lumen. Airway epithelia may initiate and amplify inflammation in response to these attacks, but under certain conditions confinement of inflammation to the airway lumen may be beneficial to the
J N Ihle et al.
Trends in biochemical sciences, 19(5), 222-227 (1994-05-01)
A variety of cytokines, lymphokines and growth factors function by interacting with receptors that are members of the cytokine receptor superfamily. These receptors share extracellular motifs and have limited similarity in their cytoplasmic domains. Although lacking catalytic domains, this family
Vu Thuy Khanh Le et al.
The Journal of general virology, 89(Pt 10), 2416-2426 (2008-09-18)
We have investigated the role of signal transducer and activator of transcription (STAT) 2 during human cytomegalovirus (HCMV) replication and found that protein levels of STAT2 are downregulated. STAT2 downregulation was observed in HCMV clinical isolates and laboratory strains with
Conor Gruber et al.
The Journal of experimental medicine, 217(5) (2020-02-25)
Type I interferonopathies are monogenic disorders characterized by enhanced type I interferon (IFN-I) cytokine activity. Inherited USP18 and ISG15 deficiencies underlie type I interferonopathies by preventing the regulation of late responses to IFN-I. Specifically, USP18, being stabilized by ISG15, sterically

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service