Skip to Content
Merck
All Photos(4)

Key Documents

05-1413

Sigma-Aldrich

Anti-CD45RA Antibody, Clone MEM 56

clone MEM 56, from mouse

Synonym(s):

CD45 antigen, Leukocyte common antigen precursor, SCID due to PTPRC deficiency, T200 glycoprotein, T200 leukocyte common antigen, Human homolog of severe combined immunodeficiency due to PTPRC deficiency, Leukocyte-common antigen, Protein tyrosine phosph

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
MEM 56, monoclonal
application:
FACS
IF
IHC
IP
WB
species reactivity:
mouse
technique(s):
flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable
citations:
2

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MEM 56, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

human, rat

technique(s)

flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PTPRC(5788)

Related Categories

General description

CD45 is a receptor-tyrosine phosphatase required for antigen-induced signaling in T-cells and B-cells; its substrates are unknown. Interestingly, it associates with several Src-family tyrosine kinases, which are inhibited by tyrosine phosphorylation in their c-termini, suggesting a mechanism for signaling by CD45.

Specificity

This antibody recognizes the 220 kDa molecular weight component of the leucocyte common antigen complex expressed on some CD4+ T lymphocytes, B lymphocytes and CD8+ lymphocytes. The CD45R+/CD4+ lymphocytes functionally exert suppressor/inducer activity in in vitro assay systems.
Antigen distribution:
B-cells (lg+) >95%
T-cells (E+) 70 + 10%
Peripheral blood lymphocytes 68 + 8%
T-cells (CD4+) 50 + 10%
Monocytes 50 + 20%
Thymocytes <5%
Granulocytes <1%

Application

Anti-CD45RA Antibody, Clone MEM 56 detects level of CD45RA & has been published & validated for use in FC, IF, IH(P), IP & WB.
Immunofluorescence: Identification of CD4+ suppressor/inducer cells, by indirect immunofluorescence staining.

Immunoprecipitation: A previous lot of this antibody detected 220 kDa molecular weight component of the leucocyte common antigen complex in immunoprecipitation.

Flow Cytometry: Purification or depletion of CD4+ suppressor/inducer cells by cell sorting or the panning techniques was performed on a previous lot.

Immunohistochemistry(paraffin): Representative testing from a previous lot.
Identification of CD45R+ lymphocytes in cryostat sections of frozen tissue and in formalin fixed wax embedded tissue.

Optimal Staining of CD45RA Monoclonal Antibody: Tonsil
Research Category
Signaling
Research Sub Category
Immunological Signaling

Quality

Routinely evaluated by Western Blot on RAW 264 lysates.

Western Blot Analysis: 1:500 dilution of this lot detected CD45RA on 10 μg of RAW 264

Target description

~220 kDa

Linkage

Replaces: 04-1102

Physical form

Format: Purified
Purified
Purified mouse monoclonal IgG2b in phosphate buffered saline containing 10 mM sodium azide and 1 mg/mL bovine serum albumin.
We recommend that each laboratory determine an optimum working titre for use in its particular application.

Storage and Stability

For use within 1 month of date of purchase store at 2-8°C, for long term storage aliquot antibody into small volumes and store at -20°C for up to 1 year from date of receipt.
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
RAW264 Cell Lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiangjun Liu et al.
Nature communications, 13(1), 1158-1158 (2022-03-05)
Cutaneous T cell lymphoma (CTCL) represents a heterogeneous group of non-Hodgkin lymphoma distinguished by the presence of clonal malignant T cells. The heterogeneity of malignant T cells and the complex tumor microenvironment remain poorly characterized. With single-cell RNA analysis and
Gaby J Ritfeld et al.
Cell transplantation, 24(11), 2209-2220 (2015-01-13)
The ability of intraspinal bone marrow stromal cell (BMSC) transplants to elicit repair is thought to result from paracrine effects by secreted trophic factors including brain-derived neurotrophic factor (BDNF). Here we used gene therapy to increase or silence BDNF production

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service