Skip to Content
Merck
All Photos(1)

Key Documents

04-281

Sigma-Aldrich

Anti-phospho-EGFR (Thr669) Antibody, clone 5F10

clone 5F10, Upstate®, from mouse

Synonym(s):

ERBB, ERBB1, epidermal growth factor receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5F10, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pThr669)

Gene Information

human ... EGFR(1956)

Specificity

Recognizes EGFR when phosphorylated at Threonine 669.

Immunogen

KLH-conjugated synthetic peptide encompassing the surrounding amino acids of Thr 669 in human EGFR.

Application

Anti-phospho-EGFR (Thr669) Antibody, clone 5F10 detects level of phospho-EGFR (Thr669) & has been published & validated for use in WB.
Research Category
Signaling
Research Sub Category
Growth Factors & Receptors

Quality

Routinely evaluated by immunoblot.

Target description

180 kDa

Physical form

100 µg of mouse monoclonal IgG1 liquid in 0.01 mg/ml PBS containing 0.09% sodium azide, PEG, and sucrose with 50 % glycerol.
Format: Purified
Thiophilic adsoption and size exclusion chromatography

Storage and Stability

6 months at -20°C from date of shipment

Analysis Note

Control
Includes PMA and pervanadate treated A431 cell lysate as a positive control

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cisplatin induces PKB/Akt activation and p38(MAPK) phosphorylation of the EGF receptor
Winograd-Katz, S E and Levitzki, A
Oncogene, 25, 7381-7390 (2006)
G J Heisermann et al.
The Journal of biological chemistry, 263(26), 13152-13158 (1988-09-15)
The epidermal growth factor (EGF) receptor is regulated by EGF-stimulated autophosphorylation and by phorbol ester-stimulated, protein kinase C (Ca2+/phospholipid-dependent enzyme) mediated phosphorylation at identified sites. The EGF receptor contains additional phosphorylation sites including a prominent phosphothreonine and several phosphoserines which
Mutational removal of the Thr669 and Ser671 phosphorylation sites alters substrate specificity and ligand-induced internalization of the epidermal growth factor receptor
Heisermann, G J, et al
The Journal of Biological Chemistry, 265, 12820-12827 (1990)
F Yan et al.
Oncogene, 36(50), 6919-6928 (2017-09-05)
Overexpression of DNMT1 and KIT is prevalent in lung cancer, yet the underlying molecular mechanisms are poorly understood. While the deregulated activation of DNMT1 or KIT has been implicated in lung cancer pathogenesis, whether and how DNMT1 and KIT orchestrate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service