Skip to Content
Merck

D-066

Dehydroepiandrosterone-D5-3-sulfate (DHEAS-D5) (2,2,3,4,4,-D5) sodium salt solution

100 μg/mL in methanol (as free sulfate), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

DHEAS-d5 (2,2,3,4,4,-D5), Dehydroepiandrosterone-d5-3-sulfate sodium salt (2,2,3,4,4,-d5)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19D5H22NaO5S
Molecular Weight:
395.50
UNSPSC Code:
41116107
MDL number:
NACRES:
NA.24
EC Number:
200-659-6
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free sulfate)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

[Na+].[2H]C1([2H])C[C@]2(C)[C@H]3CC[C@@]4(C)[C@@H](CCC4=O)[C@@H]3CC=C2C([2H])([2H])[C@@]1([2H])OS([O-])(=O)=O

InChI

1S/C19H28O5S.Na/c1-18-9-7-13(24-25(21,22)23)11-12(18)3-4-14-15-5-6-17(20)19(15,2)10-8-16(14)18;/h3,13-16H,4-11H2,1-2H3,(H,21,22,23);/q;+1/p-1/t13-,14-,15-,16-,18-,19-;/m0./s1/i7D2,11D2,13D;

InChI key

GFJWACFSUSFUOG-SJDCDROSSA-M

General description

An internal standard for DHEAS, a metabolite of dehydroepiandrosterone (DHEA). DHEAS levels are measured by LC-MS/MS for determination of excess adrenal activity as observed in adrenal cancer, certain forms of congenital adrenal hyperplasia (CAH), and polycystic ovary syndrome.

Legal Information

Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.


Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class

3 - Flammable liquids

WGK

WGK 1

Flash Point (°F)

49.5 °F - closed cup

Flash Point (°C)

9.7 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library