Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

890812P

Avanti

VSA 1

Synonym(s):

Semisynthetic Saponin Adjuvant-VSA 1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C88H145NO39
CAS Number:
Molecular Weight:
1841.08
UNSPSC Code:
12352211

Pricing and availability is not currently available.

Assay

≥99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (890812P-1MG)
pkg of 1 × 5 mg (890812P-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

vaccine development

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCNC(=O)C1OC(OC2CCC3(C)C(CCC4(C)C3CC=C3C5CC(C)(C)CCC5(CCC43C)C(=O)OC3OC(C)C(O)C(O)C3OC3OC(C)C(OC4OCC(O)C(O)C4O)C(OC4OC(CO)C(O)C(OC5OCC(O)C(O)C5O)C4O)C3O)C2(C)C=O)C(OC2OC(CO)C(O)C(O)C2O)C(OC2OC(C)C(O)C(O)C2O)C1O

General description

VSA-1 is a semisynthetic saponin adjuvant derived from the naturally occurring and readily accessible Momordica saponin, capable of stimulating antigen-specific humoral and cellular immune responses. Both VSA-1 and QS-21 significantly increased the titers of IgG and opsonic antibodies against seven selected serotypes, including serotypes 3, 14, and 19A, which are associated with most PCV13 breakthrough infections.

Application

Vaccine development

Packaging

5 mL Amber Glass Screw Cap Vial (890812P-1MG); 5 mL Amber Glass Screw Cap Vial (890812P-5MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service