Skip to Content
Merck
All Photos(1)

Key Documents

880720C

Avanti

18:0 PEG1000 PE

Avanti Research - A Croda Brand 880720C

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-1000] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

form

liquid

packaging

pkg of 1 × 2.5 mL (880720C-25mg)
pkg of 2 × 4 mL (880720C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880720C

concentration

10 mg/mL (880720C-25mg)
25 mg/mL (880720C-200mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O)=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[NH4+]

InChI key

RMRFKOYDUNAYRA-NHGDJFPISA-N

General description

The molecular weight and exact mass are averages based on the polydispersity of polyethylene glycol (PEG).[1]

Application

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-1000] (18:0 PEG1000 PE) might be used to prepare sterically stabilized liposome and study its characteristics by small-angle X-ray scattering (SAXS) method.[2] It is also suitable for use in cholesterol-free, pegylated (CF-PEG) liposomes to study its long term stability when loaded with carboplatin.[3]

Packaging

5 mL Clear Glass Sealed Ampule (880720C-200mg)
5 mL Clear Glass Sealed Ampule (880720C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z Varga et al.
Chemistry and physics of lipids, 165(4), 387-392 (2012-01-10)
Synchrotron small-angle X-ray scattering analysis of the bilayer structure of a pharmacologically relevant sterically stabilized liposome system is presented. Describing the electron density profile of the bilayer with the superposition of Gaussian functions, the contribution of the poly(ethylene glycol) (PEG)
Anumita Chaudhury et al.
International journal of pharmaceutics, 430(1-2), 167-175 (2012-04-28)
The obstacles in translating liposome formulations into marketable products could be attributed to their physical instabilities upon long-term storage as aqueous dispersions. Lyophilization is the most commonly used technique to improve physical stability of liposomes. The development of stable, lyophilized
Brian J Nablo et al.
Biophysical journal, 95(3), 1157-1164 (2008-07-23)
Nonelectrolyte polymers of poly(ethylene glycol) (PEG) were used to estimate the diameter of the ion channel formed by the Bacillus anthracis protective antigen 63 (PA(63)). Based on the ability of different molecular weight PEGs to partition into the pore and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service