Skip to Content
Merck
All Photos(1)

Key Documents

860656P

Avanti

Lyso SM (d17:1)

Avanti Research - A Croda Brand 860656P, powder

Synonym(s):

Sphingosylphosphorylcholine (C17 base)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H47N2O5P
CAS Number:
Molecular Weight:
450.59
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (860656P-10mg)
pkg of 1 × 5 mg (860656P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860656P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCC/C=C/[C@@]([H])(O)[C@@]([H])(N)COP([O-])(OCC[N+](C)(C)C)=O

InChI

1S/C22H47N2O5P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-22(25)21(23)20-29-30(26,27)28-19-18-24(2,3)4/h16-17,21-22,25H,5-15,18-20,23H2,1-4H3/b17-16+/t21-,22+/m0/s1

InChI key

AAERQLSYRRXFMA-FYBNUODKSA-N

General description

Lyso-sphingomyelin (Lyso SM) (d17:1), is an amphiphilic lysophospholipid with a long-chain sphingosine (C17 base) and a hydrophilic phosphorylcholine. Lyso SM, also known as sphingosylphosphorylcholine (SPC) is a de-acylated form of sphingomyelin, found in blood plasma.

Biochem/physiol Actions

Lyso-sphingomyelin (Lyso SM) is a vital lipid mediator, with an ability to regulate cell proliferation, migration, differentiation, metabolism and apoptosis. It exhibits various functions in cardiovascular system, immune system, central nervous system and skin. Increased concentration of Lyso SM has been observed in few disease states such as atopic dermatitis, Niemann-Pick disease (NPD) and cancer.

Packaging

5 mL Amber Glass Screw Cap Vial (860656P-10mg)
5 mL Amber Glass Screw Cap Vial (860656P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Di Ge et al.
Acta pharmacologica Sinica, 39(12), 1830-1836 (2018-07-28)
Sphingosylphosphorylcholine (SPC) is a bioactive sphingolipid in blood plasma that is metabolized from the hydrolysis of the membrane sphingolipid. SPC maintains low levels in the circulation under normal conditions, which makes studying its origin and action difficult. In recent years
Wei-Lien Chuang et al.
Molecular genetics and metabolism, 111(2), 209-211 (2014-01-15)
Niemann-Pick disease type B (NPD-B) is caused by a partial deficiency of acid sphingomyelinase activity and results in the accumulation of lysosomal sphingomyelin (SPM) predominantly in macrophages. Notably, SPM is not significantly elevated in the plasma, whole blood, or urine
Hong-Wei Yue et al.
International journal of clinical and experimental medicine, 8(8), 11913-11921 (2015-11-10)
Sphingosylphosphorylcholine (SPC) is a naturally occurring bioactive sphingolipid in blood plasma, metabolizing from the hydrolysis of the membrane sphingolipid. It has been shown to exert multifunctional role in cell physiological regulation either as an intracellular second messenger or as an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service