Skip to Content
Merck
All Photos(3)

Key Documents

860081P

Avanti

Egg Lyso PE

Avanti Research - A Croda Brand

Synonym(s):

L-α-lysophosphatidylethanolamine (Egg, Chicken)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (LPE; may contain up to 10% of the 2-LPE isomer, TLC)

form

powder

packaging

pkg of 1 × 10 mg (860081P-10mg)
pkg of 1 × 100 mg (860081P-100mg)
pkg of 1 × 25 mg (860081P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[NH3+])=O)([H])COC(CCCCCCCCCCCCCCCCC)=O

InChI

1S/C23H48NO7P/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-23(26)29-20-22(25)21-31-32(27,28)30-19-18-24/h22,25H,2-21,24H2,1H3,(H,27,28)/t22-/m1/s1

InChI key

BBYWOYAFBUOUFP-JOCHJYFZSA-N

General description

Phosphatidylethanolamine is a phospholipid and is bound to the biological membranes.[1]

Application

Egg Lyso PE is suitable for use:
  • in the activation of lysophospholipid of human transforming growth factor β1(TGF-β1)[2]
  • in determining the influence of lysoPE on binding of class II MHC molecules to peptides containing a photoreactive compound[3]
  • to check its effect on catherin mediated endocytosis(CME) in breast cancer SUM159 cells[4]

Biochem/physiol Actions

Phosphatidylethanolamine has a tendency of binding to proteins and forming a complex of phosphatidylethanolamine-binding protein (PEBP).[5]

Packaging

5 mL Amber Glass Screw Cap Vial (860081P-100mg)
5 mL Amber Glass Screw Cap Vial (860081P-10mg)
5 mL Amber Glass Screw Cap Vial (860081P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Function from structure? The crystal structure of human phosphatidylethanolamine-binding protein suggests a role in membrane signal transduction
Banfield M J, et al.
Structure, 6(10), 1245-1254 (2017)
Lysophospholipid regulates release and activation of latent TGF-beta1 from chondrocyte extracellular matrix
Gay I, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1684(1-3), 18-28 (2004)
Phosphatidylcholine?s functions beyond that of a membrane brick
Furse S and De Kroon AIPM
Molecular Membrane Biology, 32(4), 117-119 (2015)
Phospholipids enhance the binding of peptides to class II major histocompatibility molecules
Roof R W, et al.
Proceedings of the National Academy of Sciences of the USA, 87(5), 1735-1739 (1990)
Exogenous lysophospholipids with large head groups perturb clathrin-mediated endocytosis
Ailte I, et al.
Traffic, 18(3), 176-191 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service