Skip to Content
Merck
All Photos(2)

Key Documents

856715P

Avanti

18:0 Lyso PE

1-Stearoyl-2-Hydroxy-sn-Glycero-3-Phosphoethanolamine, powder

Synonym(s):

1-octadecanoyl-sn-glycero-3-phosphoethanolamine; PE(18:0/0:0); 11070

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H48NO7P
CAS Number:
Molecular Weight:
481.60
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (LPE; may contain up to 10% of the 2-LPE isomer, TLC)

form

powder

packaging

pkg of 1 × 25 mg (856715P-25mg)
pkg of 1 × 500 mg (856715P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 856715P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[NH3+])=O)([H])COC(CCCCCCCCCCCCCCCCC)=O

InChI key

BBYWOYAFBUOUFP-JOCHJYFZSA-N

Application

18:0 Lyso PE has been used as a phospholipid (PL) standard for lipid profiling by electrospray ionisation mass spectrometry (ESI-MS)/MS.[1]

Packaging

20 mL Clear Glass Screw Cap Vial (856715P-500mg)
5 mL Amber Glass Screw Cap Vial (856715P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Extraction and profiling of plant polar glycerol lipids
Liu Y and Wang X
Bio-protocol (2016)
Sidra Iqbal et al.
Plant physiology and biochemistry : PPB, 147, 101-112 (2019-12-20)
Plant phospholipase A (PLA) and C (PLC) families are least explored in terms of structure, diversity and their roles in membrane lipid remodeling under stress conditions. In this study, we performed gene family analysis, determined gene expression in different tissues
Anik Forest et al.
Journal of proteome research, 17(11), 3657-3670 (2018-09-27)
The goal of this work was to develop a label-free, comprehensive, and reproducible high-resolution liquid chromatography-mass spectrometry (LC-MS)-based untargeted lipidomic workflow using a single instrument, which could be applied to biomarker discovery in both basic and clinical studies. For this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service