Skip to Content
Merck
All Photos(1)

Key Documents

850042P

Avanti

Dicetyl phosphate

Avanti Research - A Croda Brand

Synonym(s):

Dihexadecyl phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H67O4P
CAS Number:
Molecular Weight:
546.85
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

mol wt

546.85 g/mol

packaging

pkg of 1 × 1 g (850042P-1g)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

OP(OCCCCCCCCCCCCCCCC)(OCCCCCCCCCCCCCCCC)=O

InChI

1S/C32H67O4P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-35-37(33,34)36-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-32H2,1-2H3,(H,33,34)

InChI key

RNPXCFINMKSQPQ-UHFFFAOYSA-N

Biochem/physiol Actions

Dicetyl phosphate (DCP) serves as an electric insulator. It is capable of forming a thin film on glassy carbon electrode (GCE) surface. It can be mainly used in the preparation of liposomes.

Packaging

20 mL Clear Glass Screw Cap Vial (850042P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical determination of acetaminophen using a glassy carbon electrode coated with a single-wall carbon nanotube-dicetyl phosphate film
Sun D and Zhang H
Microchimica Acta, 158(1-2), 131-136 (2007)
Fluorescence immunoassays
Immunoassay, 309-335 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service