Skip to Content
Merck
All Photos(3)

Documents

840024C

Avanti

Soy PE

Avanti Research - A Croda Brand 840024C

Synonym(s):

L-α-phosphatidylethanolamine (Soy)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840024C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840024C

concentration

10 mg/mL (840024C-25mg)

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C39H74NO8P/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-39(42)48-37(36-47-49(43,44)46-34-33-40)35-45-38(41)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2/h11,13,17-18,37H,3-10,12,14-16,19-36,40H2,1-2H3,(H,43,44)/b13-11-,18-17-/t37-/m1/s1

InChI key

HBZNVZIRJWODIB-NHCUFCNUSA-N

General description

L-α-phosphatidylethanolamine is a naturally occurring aminophospholipid in soy lecithin. Soy PE is one of the important components of biological membranes.

Application

Soy PE has been used in the preparation of liposome solutions and as a standard for the quantitative analysis of phospholipids through calibration curves.

Biochem/physiol Actions

Soy PE has an important function in maintaining the structural integrity and functionality of cells. It possesses anti-oxidative properties.

Packaging

5 mL Clear Glass Sealed Ampule (840024C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amperometric post spike feet reveal most exocytosis is via extended kiss-and-run fusion
Mellander LJ, et al.
Scientific reports, 2, 907-907 (2012)
Development and validation of a novel UPLC-ELSD method for the assessment of lipid composition of nanomedicine formulation
Varache M, et al.
International Journal of Pharmaceutics, 566, 11-23 (2019)
Identification and quantification of phosphatidylethanolamine-derived glucosylamines and aminoketoses from human erythrocytes?influence of glycation products on lipid peroxidation
Carmen MBU, et al.
Archives of Biochemistry and Biophysics, 391(2), 245-254 (2001)
Quercetin increases antioxidant activity of soy lecithin in a triolein model system
Ramadan MF
LWT--Food Science and Technology, 41(4) (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service