Skip to Content
Merck
All Photos(1)

Key Documents

W214205

Sigma-Aldrich

Benzyl cinnamate

≥98%, stabilized

Synonym(s):

3-Phenyl 2-propenoic acid benzyl ester, Benzyl 3-phenylpropenoate, Cinnamic acid benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCOOCH2C6H5
CAS Number:
Molecular Weight:
238.28
FEMA Number:
2142
Beilstein:
2051339
EC Number:
Council of Europe no.:
331
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.738
NACRES:
NA.21
grade:
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

contains

α-tocopherol, synthetic as stabilizer

bp

195-200 °C/5 mmHg (lit.)

mp

34-37 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

floral; balsamic; sweet

SMILES string

O=C(OCC1=CC=CC=C1)/C=C/C2=CC=CC=C2

InChI

1S/C16H14O2/c17-16(12-11-14-7-3-1-4-8-14)18-13-15-9-5-2-6-10-15/h1-12H,13H2/b12-11+

InChI key

NGHOLYJTSCBCGC-VAWYXSNFSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Benzylcinnamate can be found naturally in Balsam of Peru and Tolu Balsam [1]. It is a common fragrance ingredient found in various consumer products, including fine fragrances, decorative cosmetics, shampoos, and toilet soaps [2].

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

356.0 °F - closed cup

Flash Point(C)

180 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fragrance material review on benzyl cinnamate
Bhatia SP, et al.
Food And Chemical Toxicology, 45(1), S40-S48 (2007)
Cinnamein Inhibits the Induction of Nitric Oxide and Proinflammatory Cytokines in Macrophages, Microglia and Astrocytes
Pahan S, et al.
American journal of clinical and experimental immunology, 8(1), 520-529 (2023)
Osamu Ohno et al.
Bioorganic & medicinal chemistry, 16(16), 7843-7852 (2008-08-02)
Hypertension is a lifestyle-related disease which often leads to serious conditions such as heart disease and cerebral hemorrhage. Angiotensin II (Ang II) plays an important role in regulating cardiovascular homeostasis. Consequently, antagonists that block the interaction of Ang II with
Sarvesh Kumar et al.
European journal of medicinal chemistry, 46(11), 5498-5511 (2011-10-01)
In the present study, we report the design and synthesis of novel analogs of cinnamates, thiocinnamates and thionocinnamates and evaluated the potencies of these analogs to inhibit TNF-α induced ICAM-1 expression on human endothelial cells. By using whole cell-ELISA, our
Feng Li et al.
Journal of natural products, 73(4), 623-627 (2010-03-24)
Three new flavonoids, (2R,3R)-3,5-dihydroxy-7-methoxyflavanone 3-(2-methyl)butyrate (1), (7''R)-8-[1-(4'-hydroxy-3'-methoxyphenyl)prop-2-en-1-yl]chrysin (2), and (7''R)-8-[1-(4'-hydroxy-3'-methoxyphenyl)prop-2-en-1-yl]galangin (3), together with 41 known compounds (4-44) were isolated from a methanolic extract of Mexican propolis. Compounds 2 and 3 are unique natural flavones containing a 1-phenylallyl moiety. The in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service