MT51340293
Mettler-Toledo InLab® membrane kit for InLab® 605 dissolved oxygen sensor
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
51191600
NACRES:
NB.79
Recommended Products
manufacturer/tradename
Mettler-Toledo 51340293
Legal Information
InLab is a registered trademark of Mettler-Toledo, Inc.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service