M31201
4-Methylbenzylamine
97%
Synonym(s):
p-Xylylamine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
CH3C6H4CH2NH2
CAS Number:
Molecular Weight:
121.18
Beilstein:
956670
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
refractive index
n20/D 1.534 (lit.)
bp
195 °C (lit.)
mp
12-13 °C (lit.)
density
0.952 g/mL at 25 °C (lit.)
SMILES string
Cc1ccc(CN)cc1
InChI
1S/C8H11N/c1-7-2-4-8(6-9)5-3-7/h2-5H,6,9H2,1H3
InChI key
HMTSWYPNXFHGEP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
176.0 °F - closed cup
Flash Point(C)
80 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Nadeem Rezaq Janjua et al.
The Journal of investigative dermatology, 123(1), 57-61 (2004-06-12)
Recent in vitro and animal studies have reported estrogen-like activity of chemicals used in sunscreen preparations. We investigated whether the three sunscreens benzophenone-3 (BP-3), octyl-methoxycinnamate (OMC), and 3-(4-methylbenzylidene) camphor (4-MBC) were absorbed and influenced endogenous reproductive hormone levels in humans
J Ishida et al.
Analytical biochemistry, 184(1), 86-89 (1990-01-01)
A simple and highly sensitive method for the determination of beta-phenylethylamine in human plasma is investigated. The method employs high-performance liquid chromatography with fluorescence detection. beta-Phenylethylamine and p-methylbenzylamine (internal standard) in human plasma are isolated by cation-exchange chromatography on a
Renato Pirisino et al.
British journal of pharmacology, 147(2), 218-224 (2005-11-15)
1.--4-methyl benzylamine (4-MBZ; 28 microg, 231 nmol) elicits a hyperphagic response in starved mice in contrast to the hypophagia induced by the parent compound benzylamine (BZ; 33 microg, 231 nmol) or by amphetamine (AMPH, 2 mug). 2.--In mice starved for
Francesc Yraola et al.
Journal of medicinal chemistry, 49(21), 6197-6208 (2006-10-13)
Structure activity relationships for semicarbazide-sensitive amine oxidase/vascular adhesion protein-1 (SSAO/VAP-1) were studied using a library of arylalkylamine substrates, with the aim of contributing to the discovery of more efficient SSAO substrates. Experimental data were contrasted with computational docking studies, thereby
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service