GF66534958
Nickel/Iron/Molybdenum
wire, Ni80%/Fe15.5%/Mo 4.5%, length 100 m, 0.5 mm diameter, temper as drawn
Synonym(s):
NJ275140
About This Item
Recommended Products
form
wire
manufacturer/tradename
Goodfellow 665-349-58
L
100 m
diameter
0.5 mm
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service