Skip to Content
Merck
All Photos(1)

Key Documents

GF53085203

Evanohm® - precision resistance alloy, Ni75Cr20Al2.5Cu2.5

foil, 25 x 25mm, 0.025mm thickness, annealed, light tight (lt)

Synonym(s):

NI170275

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101711
NACRES:
NA.23

description

light tight (lt)

form

annealed (
quarter hard)

foil (
quarter hard)

manufacturer/tradename

Goodfellow 530-852-03

size × thickness

25 mm × 25 mm × 0.025 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Legal Information

Evanohm is a registered trademark of CRS Holdings, Inc., a subsidiary of Carpenter Technology Corporation
Evanohm is a registered trademark of CRS Holdings, Inc., a subsidiary of Carpenter Technology Corporation

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service