GF19715632
Hastelloy® C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn
100mm tube, 6.35mm outside diameter, 0.9mm wall thickness, 4.55mm inside diameter
Synonym(s):
NI147200
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
tubes
manufacturer/tradename
Goodfellow 197-156-32
L × thickness
100 mm × 0.9 mm
O.D. × I.D.
6.35 mm × 4.55 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Legal Information
Hastelloy is a registered trademark of Haynes International, Inc.
Hastelloy is a registered trademark of Haynes International, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service